The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006079.4 | NP_006070.2 | 1 - 2 | 237 | 80 |
GenBank mRNA | AF109161.1 | - | 1 - 2 | 207 | 80 |
BC004377.1 | - | 1 - 2 | 190 | 80 | |
BM763150.1 | - | 1 - 2 | 235 | 80 | |
U65093.1 | - | 1 - 2 | 190 | 80 |