Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HSPA8
Entrez Gene ID
3312
Gene Name
heat shock protein family A (Hsp70) member 8
Gene Aliases
HEL-33, HEL-S-72p, HSC54, HSC70, HSC71, HSP71, HSP73, HSPA10, LAP-1, LAP1, NIP71
Chromosome Location
Chr.11: 123057492 - 123062366 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006597.5 NP_006588.1 5 - 6 1399 111
NM_153201.3 NP_694881.1 5 - 6 1399 111
XM_011542798.1 XP_011541100.1 5 - 6 1147 111
GenBank mRNA AB034951.1 - 5 - 6 1163 111
AB073886.1 - 4 - 5 1077 111
AF352832.1 - 5 - 6 1186 111
AK096100.1 - 5 - 6 1125 111
AK129885.1 - 5 - 6 1201 111
AK222628.1 - 5 - 6 1197 111
AK222785.1 - 5 - 6 1197 111
AK292067.1 - 2 - 3 829 111
AK303935.1 - 4 - 5 970 111
BC007276.1 - 4 - 5 947 111
BC016179.1 - 5 - 6 1180 111
BC016660.1 - 5 - 6 1180 111
BC019816.2 - 5 - 6 1180 111
EU668349.1 - 5 - 6 1180 111
FJ224294.1 - 5 - 6 1200 111

View Full Product Details