The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014203.2 | NP_055018.2 | 2 - 3 | 346 | 107 |
NM_130787.2 | NP_570603.2 | 2 - 3 | 346 | 107 | |
XM_011526556.2 | XP_011524858.1 | 2 - 3 | 956 | 107 | |
XM_011526557.2 | XP_011524859.1 | 2 - 3 | 951 | 107 | |
GenBank mRNA | AK124343.1 | - | 2 - 3 | 262 | 107 |
AK302005.1 | - | 2 - 3 | 301 | 107 | |
AL136925.1 | - | 2 - 3 | 212 | 107 | |
BC014214.2 | - | 2 - 3 | 244 | 107 |