The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001007469.1 | NP_001007470.1 | 1 - 2 | 163 | 75 |
NM_014266.3 | NP_055081.1 | 1 - 2 | 163 | 75 | |
XM_017026193.1 | XP_016881682.1 | 1 - 2 | 278 | 75 | |
GenBank mRNA | AF072844.1 | - | 1 - 2 | 105 | 75 |
AF122904.1 | - | 1 - 2 | 105 | 75 | |
AF172929.1 | - | 1 - 2 | 140 | 75 | |
AF285447.1 | - | 1 - 2 | 80 | 75 | |
AL050163.1 | - | 1 - 2 | 163 | 75 | |
AY359058.1 | - | 1 - 2 | 104 | 75 | |
BC035931.1 | - | 1 - 2 | 114 | 75 | |
BC046348.1 | - | 1 - 2 | 163 | 75 | |
BC065224.1 | - | 1 - 2 | 119 | 75 |