The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128852.1 | NP_001122324.1 | 1 - 2 | 255 | 81 |
NM_001128853.1 | NP_001122325.1 | 1 - 2 | 255 | 81 | |
NM_001128854.1 | NP_001122326.1 | 1 - 2 | 255 | 81 | |
NM_015908.5 | NP_056992.4 | 1 - 2 | 255 | 81 | |
XM_017012290.1 | XP_016867779.1 | 1 - 2 | 363 | 81 | |
XM_017012291.1 | XP_016867780.1 | 1 - 2 | 363 | 81 | |
GenBank mRNA | AF248955.1 | - | 1 - 2 | 217 | 81 |
AK311728.1 | - | 1 - 2 | 230 | 81 | |
AL096723.1 | - | 1 - 2 | 209 | 81 | |
BC109117.1 | - | 1 - 2 | 149 | 81 | |
BE297447.1 | - | 1 - 2 | 241 | 81 | |
BG685713.1 | - | 1 - 2 | 222 | 81 | |
DA611314.1 | - | 1 - 2 | 256 | 81 |