The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022475.2 | NP_071920.1 | 4 - 5 | 1509 | 86 |
XM_005263178.4 | XP_005263235.1 | 4 - 5 | 1378 | 86 | |
XM_006714288.3 | XP_006714351.1 | 4 - 5 | 1378 | 86 | |
GenBank mRNA | AK291849.1 | - | 4 - 5 | 849 | 86 |
AY009317.1 | - | 4 - 5 | 1349 | 86 | |
AY009951.1 | - | 4 - 5 | 1484 | 86 | |
AY358747.1 | - | 4 - 5 | 1459 | 86 | |
BC025311.1 | - | 4 - 5 | 1335 | 86 |