Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TEX13A
Entrez Gene ID
56157
Gene Name
testis expressed 13A
Gene Aliases
-
Chromosome Location
Chr.X: 105218929 - 105220693 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291277.1 NP_001278206.1 2 - 3 539 63
NM_031274.4 NP_112564.1 2 - 3 565 63
GenBank mRNA AF285597.1 - 2 - 3 546 63
BC042547.1 - 2 - 3 520 63
BC108734.1 - 2 - 3 538 63

View Full Product Details