The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001134486.2 | NP_001127958.1 | 8 - 9 | 1792 | 73 |
NM_052942.3 | NP_443174.1 | 9 - 10 | 1900 | 73 | |
GenBank mRNA | AF288815.1 | - | 7 - 8 | 1363 | 73 |
AF328727.1 | - | 9 - 10 | 1863 | 73 | |
AF430642.1 | - | 9 - 10 | 1901 | 73 | |
AF430643.1 | - | 8 - 9 | 1781 | 73 | |
AK090479.1 | - | 7 - 8 | 4612 | 73 | |
AK315064.1 | - | 9 - 10 | 1511 | 73 | |
AY358953.1 | - | 8 - 9 | 1726 | 73 | |
BC031639.1 | - | 9 - 10 | 1887 | 73 | |
BC033761.1 | - | 5 - 6 | 1048 | 73 |