Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNTTIP1
Entrez Gene ID
116092
Gene Name
deoxynucleotidyltransferase, terminal, interacting protein 1
Gene Aliases
C20orf167, Tdif1, dJ447F3.4
Chromosome Location
Chr.20: 45791937 - 45811427 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_052951.2 NP_443183.1 2 - 3 248 60
GenBank mRNA AB035676.1 - 2 - 3 243 60
AK314003.1 - 2 - 3 217 60
BC024290.1 - 2 - 3 193 60
BG720866.1 - 2 - 3 253 60

View Full Product Details