The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053053.3 | NP_444281.1 | 7 - 8 | 1037 | 109 |
GenBank mRNA | AK290964.1 | - | 7 - 8 | 854 | 109 |
AK291922.1 | - | 7 - 8 | 977 | 109 | |
BC015401.2 | - | 7 - 8 | 862 | 109 |