The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080817.4 | NP_543007.1 | 3 - 3 | 1160 | 116 |
XM_017029414.1 | XP_016884903.1 | 2 - 2 | 1316 | 116 | |
GenBank mRNA | AJ319076.1 | - | 1 - 1 | 938 | 116 |
AK122626.1 | - | 3 - 3 | 1134 | 116 | |
AK313702.1 | - | 3 - 3 | 1135 | 116 | |
AL832460.1 | - | 3 - 3 | 1135 | 116 | |
BC070180.1 | - | 3 - 3 | 1160 | 116 |