The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133367.4 | NP_588608.1 | 1 - 2 | 123 | 119 |
GenBank mRNA | AF347029.1 | - | 1 - 2 | 113 | 119 |
AK315088.1 | - | 1 - 2 | 100 | 119 | |
BC030664.2 | - | 1 - 2 | 98 | 119 |