The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_134444.4 | NP_604393.2 | 7 - 8 | 2947 | 76 |
XM_017026344.1 | XP_016881833.1 | 5 - 6 | 2422 | 76 | |
GenBank mRNA | AF442488.1 | - | 6 - 7 | 2582 | 76 |
AF479747.1 | - | 6 - 7 | 2594 | 76 | |
AF482706.1 | - | 6 - 7 | 2592 | 76 | |
AK056688.1 | - | 5 - 6 | 2474 | 76 | |
AK315003.1 | - | 7 - 8 | 2947 | 76 | |
AY072792.1 | - | 6 - 7 | 2585 | 76 | |
BC016443.1 | - | 4 - 5 | 1054 | 76 | |
BC050326.1 | - | 7 - 8 | 2621 | 76 |