The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017442.3 | NP_059138.1 | 2 - 2 | 812 | 70 |
GenBank mRNA | AB045180.1 | - | 2 - 2 | 284 | 70 |
AB445673.1 | - | 1 - 1 | 178 | 70 | |
AF245704.1 | - | 2 - 2 | 322 | 70 | |
AF246972.1 | - | 1 - 1 | 247 | 70 | |
AF259262.1 | - | 2 - 2 | 812 | 70 | |
AF259263.1 | - | 1 - 1 | 54 | 70 | |
AY359085.1 | - | 1 - 1 | 188 | 70 | |
BC032713.1 | - | 2 - 2 | 274 | 70 |