The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005747.4 | NP_005738.4 | 1 - 2 | 62 | 69 |
GenBank mRNA | AK225620.1 | - | 1 - 2 | 58 | 69 |
BC005918.1 | - | 1 - 2 | 62 | 69 | |
BC007028.1 | - | 1 - 2 | 63 | 69 | |
BC008383.1 | - | 1 - 2 | 58 | 69 | |
BC015103.1 | - | 1 - 2 | 66 | 69 | |
D00306.1 | - | 1 - 2 | 64 | 69 |