Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COPS6
Entrez Gene ID
10980
Gene Name
COP9 signalosome subunit 6
Gene Aliases
CSN6, MOV34-34KD
Chromosome Location
Chr.7: 100088960 - 100092200 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006833.4 NP_006824.2 7 - 8 682 57
GenBank mRNA AK312024.1 - 7 - 8 673 57
BC002520.2 - 7 - 8 656 57
U70735.1 - 7 - 8 604 57

View Full Product Details