The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014061.4 | NP_054780.2 | 1 - 1 | 669 | 109 |
GenBank mRNA | AF143235.4 | - | 1 - 1 | 665 | 109 |
AF320912.1 | - | 1 - 1 | 594 | 109 | |
AK313527.1 | - | 1 - 1 | 593 | 109 | |
BC011954.1 | - | 1 - 1 | 620 | 109 |