The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321978.1 | NP_001308907.1 | 6 - 7 | 895 | 97 |
NM_001321979.1 | NP_001308908.1 | 6 - 7 | 978 | 97 | |
NM_014317.4 | NP_055132.2 | 6 - 7 | 895 | 97 | |
XM_011519437.2 | XP_011517739.1 | 3 - 4 | 1295 | 97 | |
XM_017016011.1 | XP_016871500.1 | 4 - 5 | 373 | 97 | |
GenBank mRNA | AB209763.1 | - | 5 - 6 | 2933 | 97 |
AB210838.1 | - | 6 - 7 | 653 | 97 | |
AF118395.1 | - | 5 - 6 | 515 | 97 | |
AK024802.1 | - | 6 - 7 | 660 | 97 | |
AK223414.1 | - | 6 - 7 | 660 | 97 | |
BC049211.1 | - | 6 - 7 | 605 | 97 | |
BC063635.1 | - | 6 - 7 | 623 | 97 |