The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320218.1 | NP_001307147.1 | 4 - 5 | 308 | 116 |
NM_133262.2 | NP_573569.1 | 3 - 4 | 290 | 116 | |
NM_133326.1 | NP_579872.1 | 4 - 5 | 336 | 116 | |
XM_006711163.3 | XP_006711226.1 | 2 - 3 | 295 | 116 | |
XM_011509186.2 | XP_011507488.1 | 3 - 4 | 320 | 116 | |
XM_011509187.2 | XP_011507489.1 | 3 - 4 | 361 | 116 | |
GenBank mRNA | AY039760.1 | - | 2 - 3 | 184 | 116 |
BC101129.2 | - | 3 - 4 | 226 | 116 | |
BC101130.1 | - | 2 - 3 | 210 | 116 | |
BC101131.2 | - | 3 - 4 | 256 | 116 | |
BF509031.1 | - | 1 - 2 | 116 | ||
BX114684.1 | - | 2 - 3 | 213 | 116 |