Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NRXN2
Entrez Gene ID
9379
Gene Name
neurexin 2
Gene Aliases
-
Chromosome Location
Chr.11: 64606174 - 64723188 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015080.3 NP_055895.1 20 - 21 4311 70
NM_138734.2 NP_620063.1 4 - 5 1223 70
XM_005274400.3 XP_005274457.1 19 - 20 3843 70
XM_005274401.3 XP_005274458.1 19 - 20 3843 70
XM_005274402.3 XP_005274459.1 18 - 19 3819 70
XM_011545370.1 XP_011543672.1 19 - 20 3873 70
XM_011545371.1 XP_011543673.1 19 - 20 3870 70
XM_011545373.1 XP_011543675.1 19 - 20 3852 70
XM_011545375.1 XP_011543677.1 18 - 19 3828 70
XM_011545385.1 XP_011543687.1 12 - 13 2218 70
XM_017018563.1 XP_016874052.1 18 - 19 3843 70
XM_017018564.1 XP_016874053.1 18 - 19 3840 70
XM_017018565.1 XP_016874054.1 18 - 19 3837 70
XM_017018566.1 XP_016874055.1 18 - 19 3801 70
XM_017018569.1 XP_016874058.1 17 - 18 3771 70
XM_017018570.1 XP_016874059.1 19 - 20 3873 70
XM_017018571.1 XP_016874060.1 19 - 20 3864 70
XM_017018572.1 XP_016874061.1 19 - 20 3168 70
XM_017018574.1 XP_016874063.1 18 - 19 3138 70
XM_017018575.1 XP_016874064.1 18 - 19 3135 70
XM_017018576.1 XP_016874065.1 14 - 15 2783 70
XM_017018577.1 XP_016874066.1 12 - 13 2209 70
GenBank mRNA AB035266.1 - 19 - 20 3963 70
AB177862.1 - 16 - 17 3015 70

View Full Product Details