The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286373.1 | NP_001273302.1 | 5 - 6 | 712 | 66 |
NM_001286374.1 | NP_001273303.1 | 4 - 5 | 619 | 66 | |
NM_032174.5 | NP_115550.2 | 6 - 7 | 814 | 66 | |
XM_006711572.2 | XP_006711635.1 | 6 - 7 | 689 | 66 | |
XM_011510057.2 | XP_011508359.1 | 4 - 5 | 382 | 66 | |
XM_017002481.1 | XP_016857970.1 | 6 - 7 | 652 | 66 | |
XM_017002482.1 | XP_016857971.1 | 5 - 6 | 580 | 66 | |
GenBank mRNA | AK022832.1 | - | 3 - 4 | 430 | 66 |
AK027704.1 | - | 6 - 7 | 693 | 66 | |
AK297848.1 | - | 5 - 6 | 591 | 66 | |
AL523459.3 | - | 4 - 5 | 476 | 66 | |
BC014946.1 | - | 5 - 6 | 667 | 66 | |
BX537951.1 | - | 6 - 7 | 691 | 66 |