The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144984.3 | NP_659421.1 | 2 - 3 | 504 | 71 |
GenBank mRNA | AK056299.1 | - | 2 - 3 | 481 | 71 |