The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252231.1 | NP_001239160.1 | 7 - 8 | 1549 | 66 |
NM_021982.2 | NP_068817.1 | 7 - 8 | 1549 | 66 | |
XM_006714523.2 | XP_006714586.1 | 6 - 7 | 1750 | 66 | |
XM_017008961.1 | XP_016864450.1 | 7 - 8 | 1194 | 66 | |
XM_017008963.1 | XP_016864452.1 | 6 - 7 | 1750 | 66 | |
GenBank mRNA | AJ131244.1 | - | 7 - 8 | 1212 | 66 |
AK304060.1 | - | 7 - 8 | 1538 | 66 | |
BC019341.1 | - | 7 - 8 | 1510 | 66 |