Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SCAP
Entrez Gene ID
22937
Gene Name
SREBF chaperone
Gene Aliases
-
Chromosome Location
Chr.3: 47413685 - 47477126 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001320044.1 NP_001306973.1 8 - 9 1121 127
NM_012235.3 NP_036367.2 11 - 12 1606 127
XM_005264967.1 XP_005265024.1 11 - 12 1533 127
XM_005264968.1 XP_005265025.1 8 - 9 1275 127
XM_005264971.1 XP_005265028.1 5 - 6 686 127
XM_011533501.1 XP_011531803.1 11 - 12 1633 127
XM_011533502.2 XP_011531804.1 3 - 4 282 127
XM_017005918.1 XP_016861407.1 11 - 12 1620 127
XM_017005919.1 XP_016861408.1 11 - 12 1553 127
XM_017005920.1 XP_016861409.1 11 - 12 1553 127
XM_017005921.1 XP_016861410.1 5 - 6 686 127
GenBank mRNA AK027402.1 - 5 - 6 599 127
AK291484.1 - 6 - 7 799 127
AK300311.1 - 8 - 9 1011 127
BC020987.1 - 3 - 4 327 127
BC027207.2 - 7 - 8 805 127
D83782.2 - 11 - 12 1592 127

View Full Product Details