The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001376.4 | NP_001367.2 | 60 - 61 | 11623 | 65 |
GenBank mRNA | AB002323.3 | - | 60 - 61 | 11495 | 65 |
AB290157.1 | - | 60 - 61 | 11623 | 65 | |
AL833600.1 | - | 32 - 33 | 5621 | 65 | |
BC021297.1 | - | 3 - 4 | 489 | 65 | |
BX648670.1 | - | 25 - 26 | 4225 | 65 |