Assay Design
The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.
Gene Transcripts
- Gene Symbol
- TRDV3
- Entrez Gene ID
-
28516
- Gene Name
- T cell receptor delta variable 3
- Gene Aliases
-
hDV103S1
- Chromosome Location
-
Chr.14: 22469041 - 22469614 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK301114.1 |
- |
- |
506 |
100 |
- Gene Symbol
- TRDV1
- Entrez Gene ID
-
28518
- Gene Name
- T cell receptor delta variable 1
- Gene Aliases
-
hDV101S1
- Chromosome Location
-
Chr.14: 22096050 - 22096619 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AF366516.1 |
- |
- |
144 |
100 |
View Full Product Details