The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001304548.1 | NP_001291477.1 | 3 - 4 | 583 | 104 |
NM_152632.3 | NP_689845.2 | 3 - 4 | 583 | 104 | |
XM_017029452.1 | XP_016884941.1 | 3 - 4 | 576 | 104 | |
XM_017029453.1 | XP_016884942.1 | 3 - 4 | 576 | 104 | |
GenBank mRNA | AK126295.1 | - | 3 - 4 | 549 | 104 |
BC027936.1 | - | 3 - 4 | 536 | 104 |