The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317942.1 | NP_001304871.1 | 3 - 4 | 406 | 82 |
NM_178565.4 | NP_848660.3 | 4 - 5 | 1085 | 82 | |
XM_011517018.1 | XP_011515320.1 | 4 - 5 | 1101 | 82 | |
GenBank mRNA | AK123023.1 | - | 4 - 5 | 1048 | 82 |
AK123027.1 | - | 2 - 3 | 215 | 82 | |
AY358166.1 | - | 4 - 5 | 738 | 82 | |
BC027938.1 | - | 3 - 4 | 406 | 82 |