The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001031836.2 | NP_001027006.2 | 22 - 23 | 2608 | 100 |
NR_134267.1 | 22 - 23 | 2608 | 100 | ||
XM_011544424.1 | XP_011542726.1 | 21 - 22 | 2474 | 100 | |
XM_011544426.1 | XP_011542728.1 | 22 - 23 | 2546 | 100 | |
XM_017013144.1 | XP_016868633.1 | 22 - 23 | 2462 | 100 | |
GenBank mRNA | AK302391.1 | - | 22 - 23 | 2559 | 100 |
BC028701.2 | - | 22 - 23 | 2549 | 100 |