The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136052.2 | NP_001129524.1 | 1 - 2 | 231 | 63 |
NM_001199752.2 | NP_001186681.1 | 1 - 2 | 231 | 63 | |
NM_001199753.1 | NP_001186682.1 | 1 - 2 | 231 | 63 | |
XM_011526440.2 | XP_011524742.1 | 1 - 2 | 240 | 63 | |
XM_017026265.1 | XP_016881754.1 | 1 - 2 | 223 | 63 | |
XM_017026266.1 | XP_016881755.1 | 1 - 2 | 241 | 63 | |
XM_017026267.1 | XP_016881756.1 | 1 - 2 | 230 | 63 | |
XM_017026272.1 | XP_016881761.1 | 1 - 2 | 203 | 63 | |
GenBank mRNA | AK290092.1 | - | 1 - 2 | 203 | 63 |
AK309087.1 | - | 1 - 2 | 223 | 63 | |
BC029104.1 | - | 1 - 2 | 195 | 63 |