The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001285439.1 | NP_001272368.1 | 1 - 2 | 81 | 53 |
NM_001285440.1 | NP_001272369.1 | 1 - 2 | 81 | 53 | |
NM_152756.4 | NP_689969.2 | 1 - 2 | 81 | 53 | |
XM_006714463.3 | XP_006714526.1 | 1 - 2 | 94 | 53 | |
XM_011514005.2 | XP_011512307.1 | 1 - 2 | 94 | 53 | |
XM_017009314.1 | XP_016864803.1 | 1 - 2 | 72 | 53 | |
XM_017009315.1 | XP_016864804.1 | 1 - 2 | 72 | 53 | |
GenBank mRNA | AY515854.1 | - | 1 - 2 | 77 | 53 |
BC029608.1 | - | 1 - 2 | 72 | 53 | |
BC137163.1 | - | 1 - 2 | 74 | 53 | |
BC137164.1 | - | 1 - 2 | 74 | 53 | |
BC144509.1 | - | 1 - 2 | 74 | 53 | |
BP245264.1 | - | 1 - 2 | 81 | 53 | |
CR749280.1 | - | 1 - 2 | 62 | 53 |