Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MOSPD2
Entrez Gene ID
158747
Gene Name
motile sperm domain containing 2
Gene Aliases
-
Chromosome Location
Chr.X: 14873405 - 14922166 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152581.3 NP_689794.1 1 - 2 134 87
XM_005274451.1 XP_005274508.1 1 - 2 134 87
GenBank mRNA AK315781.1 - 1 - 2 75 87
AL832230.1 - 1 - 2 96 87
BC030641.1 - 1 - 2 101 87
DA708010.1 - 1 - 2 134 87

View Full Product Details