Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SNX31
Entrez Gene ID
169166
Gene Name
sorting nexin 31
Gene Aliases
-
Chromosome Location
Chr.8: 100572882 - 100663410 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152628.3 NP_689841.3 8 - 9 835 75
XM_005250815.3 XP_005250872.1 5 - 6 474 75
XM_011516899.1 XP_011515201.1 7 - 8 812 75
XM_017013153.1 XP_016868642.1 8 - 9 835 75
XM_017013154.1 XP_016868643.1 8 - 9 835 75
XM_017013155.1 XP_016868644.1 8 - 9 835 75
XM_017013156.1 XP_016868645.1 8 - 9 835 75
XM_017013157.1 XP_016868646.1 8 - 9 835 75
XM_017013158.1 XP_016868647.1 8 - 9 835 75
XM_017013159.1 XP_016868648.1 8 - 9 835 75
XM_017013160.1 XP_016868649.1 8 - 9 835 75
XM_017013162.1 XP_016868651.1 8 - 9 835 75
XM_017013163.1 XP_016868652.1 5 - 6 915 75
GenBank mRNA AK093906.1 - 8 - 9 835 75
BC031260.1 - 5 - 6 426 75
CD106024.1 - 3 - 4 246 75

View Full Product Details