The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320652.1 | NP_001307581.1 | 2 - 3 | 352 | 87 |
NM_001763.2 | NP_001754.2 | 2 - 3 | 860 | 87 | |
GenBank mRNA | AK312945.1 | - | 2 - 3 | 596 | 87 |
BC031645.1 | - | 4 - 5 | 438 | 87 | |
DA822513.1 | - | 2 - 3 | 352 | 87 | |
M27735.1 | - | 1 - 2 | 264 | 87 | |
M28825.1 | - | 2 - 3 | 860 | 87 |