Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PIKFYVE
Entrez Gene ID
200576
Gene Name
phosphoinositide kinase, FYVE-type zinc finger containing
Gene Aliases
CFD, FAB1, HEL37, PIP5K, PIP5K3, ZFYVE29
Chromosome Location
Chr.2: 208266178 - 208358751 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178000.1 NP_001171471.1 8 - 9 1205 98
NM_015040.3 NP_055855.2 8 - 9 1205 98
NM_152671.3 NP_689884.1 6 - 7 914 98
XM_011510778.2 XP_011509080.1 9 - 10 1255 98
XM_011510779.2 XP_011509081.1 9 - 10 1188 98
XM_011510780.2 XP_011509082.1 9 - 10 1273 98
XM_011510781.2 XP_011509083.1 9 - 10 1214 98
XM_011510782.2 XP_011509084.1 9 - 10 1255 98
XM_011510783.2 XP_011509085.1 9 - 10 1255 98
XM_011510784.2 XP_011509086.1 9 - 10 1273 98
XM_011510785.2 XP_011509087.1 9 - 10 1213 98
XM_011510786.2 XP_011509088.1 7 - 8 922 98
XM_011510787.1 XP_011509089.1 7 - 8 931 98
XM_011510788.1 XP_011509090.1 6 - 7 914 98
XM_011510789.2 XP_011509091.1 5 - 6 736 98
XM_011510790.1 XP_011509092.1 7 - 8 1033 98
XM_011510792.2 XP_011509094.1 9 - 10 1255 98
XM_017003568.1 XP_016859057.1 8 - 9 1109 98
XM_017003569.1 XP_016859058.1 8 - 9 1109 98
XM_017003570.1 XP_016859059.1 6 - 7 940 98
XM_017003571.1 XP_016859060.1 5 - 6 736 98
XM_017003572.1 XP_016859061.1 7 - 8 909 98
XM_017003573.1 XP_016859062.1 4 - 5 617 98
XM_017003574.1 XP_016859063.1 3 - 4 288 98
GenBank mRNA AK127769.1 - 8 - 9 1109 99
AY457063.1 - 8 - 9 1209 98
BC017736.2 - 7 - 8 1033 98
BC032389.1 - 6 - 7 914 98
BC125052.1 - 7 - 8 936 98
BC125053.1 - 8 - 9 1194 98
EU794609.1 - 8 - 9 1209 98

View Full Product Details