The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033604.1 | NP_001028776.1 | 3 - 4 | 774 | 145 |
NM_001033605.1 | NP_001028777.1 | 3 - 4 | 774 | 145 | |
NM_014451.3 | NP_055266.2 | 3 - 4 | 774 | 145 | |
NM_198428.2 | NP_940820.1 | 3 - 4 | 774 | 145 | |
XM_005249700.3 | XP_005249757.1 | 3 - 4 | 782 | 145 | |
XM_005249701.2 | XP_005249758.1 | 3 - 4 | 782 | 145 | |
XM_011515264.2 | XP_011513566.1 | 3 - 4 | 782 | 145 | |
XM_011515265.2 | XP_011513567.1 | 3 - 4 | 581 | 145 | |
XM_011515266.2 | XP_011513568.1 | 3 - 4 | 782 | 145 | |
XM_011515267.2 | XP_011513569.1 | 3 - 4 | 782 | 145 | |
XM_011515268.1 | XP_011513570.1 | 3 - 4 | 782 | 145 | |
XM_011515270.2 | XP_011513572.1 | 3 - 4 | 782 | 145 | |
XM_017011990.1 | XP_016867479.1 | 3 - 4 | 488 | 145 | |
XM_017011991.1 | XP_016867480.1 | 3 - 4 | 490 | 145 | |
XM_017011994.1 | XP_016867483.1 | 3 - 4 | 782 | 145 | |
GenBank mRNA | AF095771.1 | - | 2 - 3 | 255 | 145 |
AK057660.1 | - | 3 - 4 | 752 | 145 | |
AK314627.1 | - | 3 - 4 | 615 | 145 | |
BC032715.1 | - | 2 - 3 | 196 | 145 | |
BC103831.2 | - | 3 - 4 | 394 | 145 | |
BX647385.1 | - | 3 - 4 | 752 | 145 |