Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM74
Entrez Gene ID
157753
Gene Name
transmembrane protein 74
Gene Aliases
NET36
Chromosome Location
Chr.8: 108606850 - 108787615 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_136411.1 3 - 4 366 112
GenBank mRNA BC033057.2 - 3 - 4 345 112

View Full Product Details