Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 108881873-108882380 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IKBKAP
Entrez Gene ID
8518
Gene Name
inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase complex-associated protein
Gene Aliases
DYS, ELP1, FD, IKAP, IKI3, TOT1
UniGene
Hs.494738
Gene Chromosome Location
Chr.9: 108867520-108934332 on Build GRCh38
Transcripts
NM_001318360.1 NM_003640.4 XM_011519136.1 XM_011519137.1 XM_017015238.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5727466 c.3223G>A p.D1075N Chr.9: 108882187-108882187 on Build GRCh38
150664 c.3285+9C>T p.? Chr.9: 108882116-108882116 on Build GRCh38
3902863 c.3241C>A p.L1081I Chr.9: 108882169-108882169 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs767800484 Chr.9: 108882164-108882164 on Build GRCh38
rs150476096 Chr.9: 108882305-108882305 on Build GRCh38
rs753410907 Chr.9: 108882197-108882197 on Build GRCh38
rs763526417 Chr.9: 108882148-108882148 on Build GRCh38
rs145411012 Chr.9: 108882352-108882352 on Build GRCh38
rs1036876048 Chr.9: 108882340-108882340 on Build GRCh38
rs578196084 Chr.9: 108881984-108881984 on Build GRCh38
rs922076976 Chr.9: 108882004-108882004 on Build GRCh38
rs376675055 Chr.9: 108882108-108882108 on Build GRCh38
rs776846807 Chr.9: 108882122-108882122 on Build GRCh38
rs759873552 Chr.9: 108882161-108882161 on Build GRCh38
rs2275495 Chr.9: 108882116-108882116 on Build GRCh38
rs997966883 Chr.9: 108882351-108882351 on Build GRCh38
rs61749203 Chr.9: 108882151-108882151 on Build GRCh38
rs755812183 Chr.9: 108882185-108882185 on Build GRCh38
rs140680615 Chr.9: 108882284-108882284 on Build GRCh38
rs748267295 Chr.9: 108882210-108882212 on Build GRCh38
rs150523182 Chr.9: 108882019-108882019 on Build GRCh38
rs967009360 Chr.9: 108881915-108881915 on Build GRCh38
rs532137930 Chr.9: 108882318-108882318 on Build GRCh38
rs993213877 Chr.9: 108882252-108882252 on Build GRCh38
rs998227927 Chr.9: 108882042-108882042 on Build GRCh38
rs1015755882 Chr.9: 108881983-108881983 on Build GRCh38
rs745735611 Chr.9: 108882114-108882114 on Build GRCh38
rs978326962 Chr.9: 108881953-108881953 on Build GRCh38
rs374434020 Chr.9: 108882126-108882126 on Build GRCh38
rs781491102 Chr.9: 108881960-108881960 on Build GRCh38
rs16913636 Chr.9: 108882259-108882259 on Build GRCh38
rs888264912 Chr.9: 108881914-108881914 on Build GRCh38
rs146440397 Chr.9: 108882130-108882130 on Build GRCh38
rs1011473258 Chr.9: 108882224-108882224 on Build GRCh38
rs955106226 Chr.9: 108882269-108882269 on Build GRCh38
rs546846764 Chr.9: 108882290-108882290 on Build GRCh38
rs749199634 Chr.9: 108882091-108882091 on Build GRCh38
rs150595103 Chr.9: 108882127-108882127 on Build GRCh38
rs752344797 Chr.9: 108882174-108882174 on Build GRCh38
rs575321959 Chr.9: 108881895-108881895 on Build GRCh38
rs200527819 Chr.9: 108882098-108882098 on Build GRCh38
rs187191108 Chr.9: 108882295-108882295 on Build GRCh38
rs139864506 Chr.9: 108882229-108882229 on Build GRCh38
rs143315501 Chr.9: 108882256-108882256 on Build GRCh38
rs777231662 Chr.9: 108882196-108882196 on Build GRCh38
rs372907186 Chr.9: 108882203-108882203 on Build GRCh38
rs773477699 Chr.9: 108882134-108882134 on Build GRCh38
rs777591864 Chr.9: 108882322-108882322 on Build GRCh38
rs560928595 Chr.9: 108882137-108882137 on Build GRCh38
rs7046977 Chr.9: 108881963-108881963 on Build GRCh38
rs200941360 Chr.9: 108882129-108882129 on Build GRCh38
rs745599497 Chr.9: 108882023-108882023 on Build GRCh38
rs551466078 Chr.9: 108882325-108882325 on Build GRCh38
rs924656000 Chr.9: 108882247-108882247 on Build GRCh38
rs896859460 Chr.9: 108882343-108882343 on Build GRCh38
rs956149807 Chr.9: 108882251-108882251 on Build GRCh38
rs951961092 Chr.9: 108882090-108882090 on Build GRCh38

View Full Product Details