Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CGGBP1
Entrez Gene ID
8545
Gene Name
CGG triplet repeat binding protein 1
Gene Aliases
CGGBP, p20-CGGBP
Chromosome Location
Chr.3: 88051950 - 88149866 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195308.1 NP_001182237.1 2 - 3 259 65

View Full Product Details