Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 73160147-73160666 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANXA1
Entrez Gene ID
301
Gene Name
annexin A1
Gene Aliases
ANX1, LPC1
UniGene
Hs.494173
Gene Chromosome Location
Chr.9: 73151731-73170393 on Build GRCh38
Transcripts
NM_000700.2 XM_011518609.1 XM_017014657.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6054046 c.397G>T p.E133* Chr.9: 73160356-73160356 on Build GRCh38
6240480 c.379A>G p.M127V Chr.9: 73160371-73160371 on Build GRCh38
1462934 c.348G>A p.A116A Chr.9: 73160340-73160340 on Build GRCh38
3382566 c.381G>A p.A127A Chr.9: 73160340-73160340 on Build GRCh38
5876368 c.337G>A p.G113S Chr.9: 73160296-73160296 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753709791 Chr.9: 73160270-73160270 on Build GRCh38
rs753397823 Chr.9: 73160627-73160627 on Build GRCh38
rs992725916 Chr.9: 73160592-73160592 on Build GRCh38
rs769227185 Chr.9: 73160332-73160332 on Build GRCh38
rs1053093162 Chr.9: 73160219-73160219 on Build GRCh38
rs781651862 Chr.9: 73160349-73160363 on Build GRCh38
rs973533196 Chr.9: 73160464-73160464 on Build GRCh38
rs368581314 Chr.9: 73160383-73160383 on Build GRCh38
rs752630316 Chr.9: 73160237-73160237 on Build GRCh38
rs948432940 Chr.9: 73160182-73160182 on Build GRCh38
rs758676101 Chr.9: 73160385-73160385 on Build GRCh38
rs921889371 Chr.9: 73160482-73160482 on Build GRCh38
rs187516595 Chr.9: 73160422-73160422 on Build GRCh38
rs574714533 Chr.9: 73160549-73160549 on Build GRCh38
rs567460704 Chr.9: 73160463-73160463 on Build GRCh38
rs866217374 Chr.9: 73160375-73160375 on Build GRCh38
rs114169835 Chr.9: 73160415-73160415 on Build GRCh38
rs944302611 Chr.9: 73160527-73160527 on Build GRCh38
rs558619877 Chr.9: 73160577-73160577 on Build GRCh38
rs879233173 Chr.9: 73160216-73160216 on Build GRCh38
rs1050305 Chr.9: 73160319-73160319 on Build GRCh38
rs527409000 Chr.9: 73160178-73160178 on Build GRCh38
rs772048557 Chr.9: 73160476-73160476 on Build GRCh38
rs996876713 Chr.9: 73160343-73160343 on Build GRCh38
rs267602264 Chr.9: 73160362-73160362 on Build GRCh38
rs1042778263 Chr.9: 73160186-73160186 on Build GRCh38
rs576077015 Chr.9: 73160564-73160564 on Build GRCh38
rs996888094 Chr.9: 73160563-73160563 on Build GRCh38
rs987504264 Chr.9: 73160526-73160526 on Build GRCh38
rs960268609 Chr.9: 73160588-73160588 on Build GRCh38
rs781710936 Chr.9: 73160413-73160413 on Build GRCh38
rs780151305 Chr.9: 73160311-73160311 on Build GRCh38
rs747122086 Chr.9: 73160397-73160397 on Build GRCh38
rs536114432 Chr.9: 73160528-73160528 on Build GRCh38
rs541248294 Chr.9: 73160179-73160179 on Build GRCh38
rs1024853465 Chr.9: 73160395-73160395 on Build GRCh38
rs932202313 Chr.9: 73160239-73160239 on Build GRCh38
rs780088641 Chr.9: 73160396-73160396 on Build GRCh38
rs559382248 Chr.9: 73160250-73160251 on Build GRCh38
rs201927957 Chr.9: 73160423-73160423 on Build GRCh38
rs750714155 Chr.9: 73160384-73160384 on Build GRCh38
rs1052020700 Chr.9: 73160246-73160246 on Build GRCh38
rs762166531 Chr.9: 73160338-73160338 on Build GRCh38
rs768549264 Chr.9: 73160409-73160409 on Build GRCh38
rs540630681 Chr.9: 73160469-73160469 on Build GRCh38
rs770196300 Chr.9: 73160420-73160420 on Build GRCh38
rs770135039 Chr.9: 73160339-73160339 on Build GRCh38
rs753762644 Chr.9: 73160373-73160373 on Build GRCh38
rs534630933 Chr.9: 73160295-73160295 on Build GRCh38
rs34902839 Chr.9: 73160425-73160425 on Build GRCh38
rs757876850 Chr.9: 73160298-73160299 on Build GRCh38
rs1011858234 Chr.9: 73160580-73160580 on Build GRCh38
rs759209422 Chr.9: 73160345-73160345 on Build GRCh38
rs892736960 Chr.9: 73160516-73160516 on Build GRCh38
rs767043877 Chr.9: 73160234-73160234 on Build GRCh38
rs542717096 Chr.9: 73160250-73160250 on Build GRCh38
rs757063208 Chr.9: 73160293-73160293 on Build GRCh38
rs772091621 Chr.9: 73160310-73160310 on Build GRCh38
rs779281935 Chr.9: 73160294-73160294 on Build GRCh38
rs899973995 Chr.9: 73160561-73160561 on Build GRCh38
rs993092983 Chr.9: 73160403-73160403 on Build GRCh38
rs746085433 Chr.9: 73160301-73160301 on Build GRCh38
rs1001121851 Chr.9: 73160206-73160206 on Build GRCh38
rs535162637 Chr.9: 73160447-73160447 on Build GRCh38
rs752285742 Chr.9: 73160363-73160363 on Build GRCh38
rs892649931 Chr.9: 73160257-73160257 on Build GRCh38
rs777534277 Chr.9: 73160266-73160266 on Build GRCh38
rs1022807917 Chr.9: 73160599-73160599 on Build GRCh38
rs758401357 Chr.9: 73160249-73160250 on Build GRCh38
rs774051268 Chr.9: 73160340-73160340 on Build GRCh38
rs17574438 Chr.9: 73160200-73160200 on Build GRCh38
rs1046087737 Chr.9: 73160582-73160582 on Build GRCh38
rs550451804 Chr.9: 73160371-73160371 on Build GRCh38
rs562768663 Chr.9: 73160287-73160287 on Build GRCh38
rs757186022 Chr.9: 73160380-73160380 on Build GRCh38
rs747492937 Chr.9: 73160286-73160287 on Build GRCh38
rs10119654 Chr.9: 73160567-73160567 on Build GRCh38
rs553685193 Chr.9: 73160461-73160461 on Build GRCh38
rs766978843 Chr.9: 73160347-73160347 on Build GRCh38
rs556510076 Chr.9: 73160531-73160531 on Build GRCh38
rs373805146 Chr.9: 73160218-73160218 on Build GRCh38
rs1015577202 Chr.9: 73160584-73160584 on Build GRCh38
rs756050061 Chr.9: 73160263-73160263 on Build GRCh38
rs763707953 Chr.9: 73160579-73160579 on Build GRCh38
rs759031678 Chr.9: 73160221-73160221 on Build GRCh38
rs894827740 Chr.9: 73160366-73160366 on Build GRCh38

View Full Product Details