Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZBTB43
Entrez Gene ID
23099
Gene Name
zinc finger and BTB domain containing 43
Gene Aliases
ZBTB22B, ZNF-X, ZNF297B
Chromosome Location
Chr.9: 126804072 - 126838210 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014007.3 NP_054726.1 2 - 3 250 75
XM_005251835.3 XP_005251892.1 2 - 3 571 75
XM_011518409.1 XP_011516711.1 3 - 4 263 75
GenBank mRNA AF049907.1 - 2 - 3 230 75
AK026123.1 - 2 - 3 196 75
BC008828.2 - 2 - 3 174 75

View Full Product Details