Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- TBCD
- Entrez Gene ID
-
6904
- Gene Name
- tubulin folding cofactor D
- Gene Aliases
-
SSD-1, tfcD
- Chromosome Location
-
Chr.17: 82752048 - 82943186 on Build GRCh38
View Full Product Details