The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003907.2 | NP_003898.2 | 12 - 13 | 2104 | 76 |
XM_011513265.1 | XP_011511567.1 | 8 - 9 | 1019 | 76 | |
XM_011513266.2 | XP_011511568.1 | 11 - 12 | 1580 | 76 | |
GenBank mRNA | AK091646.1 | - | 12 - 13 | 1845 | 76 |
AK310207.1 | - | 12 - 13 | 1750 | 76 | |
AL832179.1 | - | 12 - 13 | 1764 | 76 | |
BC013590.1 | - | 12 - 13 | 1790 | 76 | |
U23028.1 | - | 11 - 12 | 1500 | 76 |