The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042663.1 | NP_001036128.1 | 18 - 19 | 2207 | 75 |
NM_001042664.1 | NP_001036129.1 | 17 - 18 | 2161 | 75 | |
NM_001042665.1 | NP_001036130.1 | 17 - 18 | 2144 | 75 | |
NM_001265592.1 | NP_001252521.1 | 18 - 19 | 2240 | 75 | |
NM_001265593.1 | NP_001252522.1 | 17 - 18 | 2171 | 75 | |
NM_001265594.1 | NP_001252523.1 | 17 - 18 | 2175 | 75 | |
NM_020631.4 | NP_065682.2 | 17 - 18 | 2090 | 75 | |
NM_198681.3 | NP_941374.2 | 18 - 19 | 2719 | 75 | |
GenBank mRNA | AB018263.2 | - | 18 - 19 | 2195 | 75 |
AB097001.1 | - | 16 - 17 | 1936 | 75 | |
AK024676.1 | - | 17 - 18 | 2089 | 75 | |
AK091201.1 | - | 5 - 6 | 1140 | 75 | |
AK096347.1 | - | 18 - 19 | 2667 | 75 | |
AK125550.1 | - | 18 - 19 | 2173 | 75 | |
AK131074.1 | - | 15 - 16 | 2414 | 75 | |
AK294875.1 | - | 18 - 19 | 2240 | 75 | |
AK299523.1 | - | 17 - 18 | 2171 | 75 | |
AK301239.1 | - | 11 - 12 | 1170 | 75 | |
BC015231.1 | - | 17 - 18 | 2151 | 75 | |
BC036671.2 | - | 18 - 19 | 2720 | 75 |