The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002218.4 | NP_002209.2 | 14 - 15 | 1925 | 92 |
GenBank mRNA | BC136393.1 | - | 14 - 15 | 1897 | 92 |
D38595.1 | - | 14 - 15 | 1899 | 92 |