Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHMP2B
Entrez Gene ID
25978
Gene Name
charged multivesicular body protein 2B
Gene Aliases
ALS17, CHMP2.5, DMT1, VPS2-2, VPS2B
Chromosome Location
Chr.3: 87227263 - 87255548 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014043.3 NP_054762.2 1 - 2 297 72
GenBank mRNA AF151842.1 - 1 - 2 256 72
AK002180.1 - 1 - 2 275 72
AK222654.1 - 1 - 2 257 72
AK316585.1 - 1 - 2 261 72
AL080122.1 - 1 - 2 270 72
BC001553.1 - 1 - 2 113 72
BI821848.1 - 1 - 2 274 72
CR533456.1 - 1 - 2 37 72

View Full Product Details