The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021953.3 | NP_068772.2 | 5 - 6 | 1261 | 77 |
NM_202002.2 | NP_973731.1 | 5 - 6 | 1261 | 77 | |
XM_005253676.3 | XP_005253733.1 | 5 - 6 | 1261 | 77 | |
XM_011520930.2 | XP_011519232.1 | 5 - 6 | 1258 | 77 | |
XM_011520932.1 | XP_011519234.1 | 5 - 6 | 1261 | 77 | |
XM_011520933.1 | XP_011519235.1 | 5 - 6 | 1261 | 77 | |
XM_011520934.2 | XP_011519236.1 | 5 - 6 | 1261 | 77 | |
GenBank mRNA | AY542306.1 | - | 4 - 5 | 1029 | 77 |
BC012863.1 | - | 4 - 5 | 997 | 77 | |
BT006986.1 | - | 4 - 5 | 978 | 77 | |
U74612.1 | - | 5 - 6 | 1092 | 77 | |
U83113.1 | - | 5 - 6 | 1047 | 77 |