The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301065.1 | NP_001287994.1 | 1 - 2 | 133 | 119 |
NM_007180.2 | NP_009111.2 | 1 - 2 | 133 | 119 | |
GenBank mRNA | AB000824.1 | - | 1 - 2 | 143 | 119 |
AK223140.1 | - | 1 - 2 | 107 | 119 | |
AK223143.1 | - | 1 - 2 | 111 | 119 | |
AK309565.1 | - | 1 - 2 | 107 | 119 | |
BC109206.2 | - | 1 - 2 | 127 | 119 |