The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286164.1 | NP_001273093.1 | 7 - 8 | 1667 | 71 |
NM_004319.2 | NP_004310.1 | 7 - 8 | 1667 | 71 | |
NM_207108.2 | NP_996991.1 | 7 - 8 | 1667 | 71 | |
GenBank mRNA | AB006627.1 | - | 7 - 8 | 1465 | 71 |
AK295126.1 | - | 7 - 8 | 1619 | 71 | |
AK307820.1 | - | 7 - 8 | 1578 | 71 | |
AK309094.1 | - | 7 - 8 | 1736 | 71 | |
BC142697.1 | - | 7 - 8 | 1446 | 71 | |
BC146795.1 | - | 7 - 8 | 1465 | 71 |