Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ASTN1
Entrez Gene ID
460
Gene Name
astrotactin 1
Gene Aliases
ASTN
Chromosome Location
Chr.1: 176855118 - 177164904 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286164.1 NP_001273093.1 19 - 20 3452 78
NM_004319.2 NP_004310.1 19 - 20 3452 78
NM_207108.2 NP_996991.1 19 - 20 3452 78
XM_017001340.1 XP_016856829.1 19 - 20 3426 78
XM_017001341.1 XP_016856830.1 19 - 20 3426 78
GenBank mRNA AB006627.1 - 19 - 20 3250 78
AK295126.1 - 19 - 20 3404 78
BC015438.1 - 7 - 8 1096 78
BC142697.1 - 19 - 20 3231 78
BC146795.1 - 19 - 20 3250 78

View Full Product Details